Learn More
Buerkle™ SteriBag™ Sampling Bags
BIOPHARMA
Product Code. 10574021 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
60 mL
150 mL
450 mL
650 mL
800 mL
1650 mL
Product Type:
Sample Bag
Sample Bag with Labeling Area
Unit Size:
Pack of 250
Pack of 500
Product Code. Capacity (Metric) Product Type unitSize
10574021 150 mL Sample Bag with Labeling Area Pack of 500
10389720 60 mL Sample Bag with Labeling Area Pack of 500
15306297 1650 mL Sample Bag Pack of 250
10135550 60 mL Sample Bag Pack of 500
10607871 150 mL Sample Bag Pack of 500
10625951 450 mL Sample Bag with Labeling Area Pack of 500
10490951 450 mL Sample Bag Pack of 500
10772141 650 mL Sample Bag with Labeling Area Pack of 500
10658251 650 mL Sample Bag Pack of 500
10584021 800 mL Sample Bag with Labeling Area Pack of 500
10544021 800 mL Sample Bag Pack of 500
10724784 1650 mL Sample Bag with Labeling Area Pack of 250
12 options
Product Code. 10574021
Brand: Buerkle™ 53440181
Only null left
Add to basket
Add to basket
Made of polyethylene and are exceptionally tearproof and leakproof. Buerkle™ SteriBags™ Sampling Bags are sterilized with tear-open closure and can be labelled.
SteriBag sterilized sampling bags are ideal for samples in chemistry, pharmaceuticals, medicine, veterinary science and foodstuffs. The bags are made of polyethylene and are therefore exceptionally tearproof and leakproof.
  • Sterilized
  • Can be labelled
  • Leakproof
  • Save space
  • Transparent
  • With practical tear-open closure
  • Proven millions of times
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 150 mL
Closure Type Tear-open
Color Clear
For Use With (Application) Ideal for Samples in Chemistry, Pharmaceuticals, Medicine, Veterinary Science and Foodstuffs
Material Polyethylene
Sterility Sterile
No. per Pack 500
Permeability Leakproof
Quantity 500/Pk.
Dimensions (L x W) 178 x 76 mm
Length (Metric) 178 mm
Width (Metric) 76 mm
Product Type Sample Bag with Labeling Area