Learn More
Buerkle™ Polyethylene SteriBag™ Sampling Bag
Shop All Bürkle Products
Click to view available options
Dimensions (L x W):
305 x 406 mm
Unit Size:
Pack of 250
Description
- Sterilised
- Can be labelled
- Leakproof
- Save space
- Transparent
- With practical closure to tear-open
- Proven millions of times
Specifications
Specifications
| Material | Polyethylene |
| Sterility | Sterile |
| Quantity | 250/Pk. |
| Dimensions (L x W) | 305 x 406 mm |
| Length (Metric) | 305 mm |
| Width (Metric) | 406 mm |
| Product Type | Sample Bag |