Learn More
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Non-sterile
GREENER_CHOICE
Product Code. 15883895 Shop All Corning Products
Click to view available options
Volume (Metric):
0.05 mL
0.5 mL
1.25 mL
2.5 mL
5 mL
12.5 mL
25 mL
50 mL
Unit Size:
Pack of 100
Pack of 25
Product Code. 15883895
Brand: Corning™ 6624
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Non-sterile provide precise, positive-displacement dispensing of viscous or volatile liquids.
  • Ideal for viscous, dense and volatile liquides
  • Minimum increments printed on the syringe itself
  • Accurately deliver selected dose
  • Made from high quality polypropylene and polyethylene
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Certifications/Compliance ISO8655
Material Polypropylene
Sterility Non-sterile
For Use With (Application) Dispensing of viscous and volatile liquids
Quantity 100/Pk.
Volume (Metric) 5 mL