Learn More
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Non-sterile
GREENER_CHOICE
Product Code. 15843895 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
0.05 mL
0.5 mL
1.25 mL
2.5 mL
5 mL
12.5 mL
25 mL
50 mL
Unit Size:
Pack of 100
Pack of 25
Product Code. Volume (Metric) unitSize
15843895 0.05 mL Pack of 100
15863895 1.25 mL Pack of 100
15873895 2.5 mL Pack of 100
15853895 0.5 mL Pack of 100
15803905 25 mL Pack of 25
15883895 5 mL Pack of 100
15893895 12.5 mL Pack of 100
15813905 50 mL Pack of 25
8 options
Product Code. 15843895
Brand: Corning™ 6620
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Non-sterile provide precise, positive-displacement dispensing of viscous or volatile liquids.
  • Ideal for viscous, dense and volatile liquides
  • Minimum increments printed on the syringe itself
  • Accurately deliver selected dose
  • Made from high quality polypropylene and polyethylene
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Certifications/Compliance ISO8655
Material Polypropylene
Sterility Non-sterile
For Use With (Application) Dispensing of viscous and volatile liquids
Quantity 100/Pk.
Volume (Metric) 0.05 mL