Learn More
R&D Systems™ StemXVivo EMT Inducing Media Supplement (100X)
Product Code. 18441320
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18441320 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18441320 Supplier R&D Systems™ Supplier No. CCM017
  • ONLINE EXCLUSIVE PRICE
    6805.00 NOK
    6435.45 NOK / 1mL
    Save 369.55 NOK
    5% Off
View all promotions
 Restricted Item
In Stock
To reliably induce epithelial to mesenchymal transition (EMT) in vitro
Reliable induction of EMT is useful for understanding the biology of EMT and for identifying and evaluating novel prognostic markers and therapeutics for fibrotic diseases and metastasis. However, the ability to induce EMT in vitro typically requires TGF-beta stimulation or labor-intensive genetic modification. Moreover, although TGF-beta is necessary to induce EMT, it is not sufficient in all cell types.
  • Drives EMT in 5 days to ensure efficient use of time and reagents
  • Provides a straightforward method to induce EMT in vitro
  • Is completely defined to reduce experimental variability
  • May induce EMT in cells resistant to TGF-beta stimulation alone
  • Avoids time-consuming genetic modifications
TRUSTED_SUSTAINABILITY
Specifications
Product Type Stem Cell Supplement Media
Quantity 1 mL
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.