Learn More
Gibco™ StemPro™ BM Mesenchymal Stem Cells
Product Code. 15373816
Click to view available options
Quantity:
1 x 106 cells
Unit Size:
Each
Product Code. 15373816
Brand: Gibco™ A15652
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
StemPro™ BM Mesenchymal Stem Cells
StemPro™ BM Mesenchymal Stem Cells are cryopreserved human bone marrow mesenchymal stem cells (BM MSCs). These Off-the-Shelf cells are manufactured to meet Good Manufacturing Practice (GMP) manufacturing standards in a California-licensed facility and available for Research Use Only. They are isolated and expanded in culture to passage 4 under a low oxygen (ischemia tolerant (it)) proprietary clinical-scale manufacturing process using Bovine Growth Serum (BGS)-supplemented media and cryopreserved in an animal origin free cryopreservation medium. The cells are supplied at 1x106 and 5x106 viable cells/vial. A Certificate of Analysis is available for each lot which documents the quality control specifications, test results and donor information.
StemPro™ BM Mesenchymal Stem Cells allow you to:
  • Minimize assay variability with itMSCs manufactured in compliance with GMP manufacturing standards
  • Improve functionality studies with immature, high potency MSCs
  • No observed tumorigenicity and toxicity in GLP-compliant animal studies
  • Accommodate experiments requiring one large lot of MSCs
  • Minimize your chance of assay failure when using tested cells, reagents and protocols
    Immature, High Potency, Low Oxygen-Cultured MSCs
    StemPro™ BM Mesenchymal Stem Cells are derived from human bone marrow aspirates from qualified, traceable donors. The cells are isolated, cultured and expanded in compliance with GMP manufacturing standards in a California-licensed facility using a proprietary clinical-scale Reduced Oxygen Tension manufacturing process. Cells manufactured in reduced oxygen tension environments result in higher cell yields of highly potent immature stem cells compared to cells expanded in normal oxygen culture conditions.
    Proven Non-toxic Profiles
    Cells have demonstrated to be non-toxic and non-tumorigenic in GLP animal studies, which were performed according to FDA guidelines.
    A large lot can be made available for those requiring assay standardization. Protocols using tested reagents will be provided to help minimize your chance of assay failure when using reagents and cells that have not been evaluated to work together. View our tested media and reagents.
    High Post-Cryopreservation Viability
    StemPro™ Mesenchymal Stem Cells are supplied in CryoStor™ cryopreservation medium at 1x106 and 5x106 viable cells/vial. This medium is animal origin free and shown to support high cell viabilities post-thawing. Controlled processes are used for cryopreservation, handling and shipping to help minimize risk of adverse effects to the quality of the cells.
    Made in USA by Stemedica for Thermo Fisher Scientific.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Cell Passage Number Passage 4
    Content And Storage 1 vial. Remove from foil pouch and store in liquid nitrogen.
    Age Adult
    Cell Type Stem Cell (Mesenchymal)
    Culture Type Adherent Cell Culture
    Form Cryopreserved
    Species Human
    Tissue Bone Marrow
    Donor Attributes Normal
    Media Recommendation Serum-Supplemented or Serum-Free Media
    No. of Cells 1 x 106 Cells
    Product Line StemPro
    Quantity 1 x 106 cells