missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies STEMdiff™ Forebrain Neuron Differentiation Kit
Description
- The STEMdiff™ Forebrain Neuron Differentiation Kit is used in conjunction with the STEMdiff™ Forebrain Neuron Maturation Kit (Catalog #08605) to generate a mixed population of forebrain-type (FOXG1-positive) neurons from neural progenitor cells derived from human pluripotent stem cells
- This kit is optimized to work with STEMdiff™ SMADi Neural Induction Kit, which supplies the appropriate neural progenitor cells
- Neurons derived using these products are versatile tools for modeling human neurological development and disease, drug screening, toxicity testing, and cell therapy validation
Specifications
Specifications
| Cell Type | Neural Cells, PSC-Derived, Neural Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture, Differentiation |
| Quantity | 1 Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?