missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies STEMdiff™ Endothelial Differentiation Kit

Product Code. 17189931
Encompass
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17189931

Brand: STEMCELL Technologies 08005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Efficient differentiation of human pluripotent stem cells to endothelial cells

  • STEMdiff™ Endothelial Differentiation Kit (Catalog #08005) includes attachment substrate, animal component-free (ACF) endothelial induction medium, and endothelial expansion medium
  • It is optimized for the differentiation of human pluripotent stem cells (hPSCs) maintained in mTeSR™1 (Catalog #85850), mTeSR™ Plus (Catalog #05825), or TeSR™-E8™ (Catalog #05990) on Corning™ Matrigel™ to endothelial-like cells
  • The kit is designed to be used immediately after early mesoderm induction with STEMdiff™ Mesoderm Induction Medium (Catalog #05220), available for purchase separately

Specifications

Cell Type Endothelial Cells
For Use With (Application) Differentiation
Quantity 1 Kit
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.