Learn More
STEMCELL Technologies STEMdiff™ Definitive Endoderm Kit (TeSR™-E8™ Optimized)
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Description
- STEMdiff™ Definitive Endoderm Kit (TeSR™-E8™ Optimized) is a complete, serum- and animal component-free medium and supplement kit that supports highly efficient differentiation of human ES cells and iPS cells that have been maintained in TeSR™-E8™ to definitive endoderm
- Cells differentiated using the STEMdiff™ Definitive Endoderm Kit (TeSR™-E8™ Optimized) express high levels of endoderm markers, including CD184 (CXCR4) and SOX17
- When used in combination with Vitronectin XF™, the STEMdiff™ Definitive Endoderm Kit (TeSR™-E8™ Optimized) provides a completely xeno-free workflow for the generation of multipotent definitive endoderm
- Optimized for differentiation of cells maintained in TeSR™-E8™
Specifications
Specifications
| Cell Type | Airway Cells, Endoderm, PSC-Derived, Hepatic Cells, Intestinal Cells, Pancreatic Cells, Pluripotent Stem Cells |
| For Use With (Application) | Cell Culture, Differentiation |
| Quantity | 1 Kit |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.