Learn More
STEMCELL Technologies STEMdiff™ Cerebral Organoid Maturation Kit
Product Code. 16648927
Encompass
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Product Code. 16648927
Brand: STEMCELL Technologies 08571
Culture medium kit for extended maturation of human cerebral organoids
  • Generate self-organized, pluripotent stem cell (PSC)-derived neural organoids with a cellular composition and structural organization representative of the developing human brain
  • This simple, four-stage protocol is based on the formulation published by Lancaster et al. (Lancaster MA et al. Nature, 2013 and Lancaster MA et al. Science, 2014) to more reliably generate cerebral organoids
  • Beginning with an embryoid body (EB) formation step followed by expansion of neuroepithelia, organoids generated using STEMdiff Cerebral Organoid Kit feature cortical-like regions including the ventricular zone, outer subventricular zone, intermediate zone, and cortical plate
  • For extended culture periods (> 40 days), the components required for maturation can be purchased as STEMdiff Cerebral Organoid Maturation Kit (Catalog #08571)
Specifications
Cell Type Neural Cells, PSC-Derived, Neural Stem and Progenitor Cells, Pluripotent Stem Cells
For Use With (Application) Cell Culture, Differentiation, Organoid Culture
Quantity 1 Kit
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.