Learn More
Cole-Parmer™ Integrity 10, RS-200, and RS-250 Reaction Station Accessories
Product Code. 15487844
Change view
Click to view available options
Product Type:
Reducing Sleeve
Reflux Head
Small Volume Test Tube (3 mL)
Small Volume Test Tubes (1 mL)
Unit Size:
Each
Product Code. Product Type unitSize
15487844 Small Volume Test Tube (3 mL) Each
15487854 Reducing Sleeve Each
15447854 Small Volume Test Tubes (1 mL) Each
11806752 Reflux Head Each
4 options
Product Code. 15487844
Brand: Cole-Parmer™ 3663083
Only null left
Add to basket
Add to basket
Keep your reaction station running smoothly with these parts and accessories.
  • Suitable for use with Cole-Parmer™ Integrity 10, RS-200, or RS-250 series reaction stations
  • Choose from a wide range of accessories suitable for your reaction station including reaction vessels, probes, and software
Specifications
Product Type Small Volume Test Tube (3 mL)
For Use With (Equipment) Integrity 10, RS-200, and RS-250 Reaction Stations
Material Glass