Learn More
Bode Chemie™ Stellisept™ med Antimicrobial Washing Lotion
Catalog No. 10785924
Ansicht ändern
Click to view available options
For Use With (Application):
For antimicrobial body cleansing (in case of MRSA/ORSA, VRE) including shampooing. Through selected ingredients and the alcohol-free formulation the product possesses a considerably better skin compatibility than conventional decontamination preparations.
Packungsgröße:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Packungsgröße
10785924 For antimicrobial body cleansing (in case of MRSA/ORSA, VRE) including shampooing. Through selected ingredients and the alcohol-free formulation the product possesses a considerably better skin compatibility than conventional decontamination preparations.
1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Catalog No. 10785924 Lieferant Bode Chemie™ Lieferanten-Nr. 9804981
  • ONLINE EXCLUSIVE PRICE
    189.57 NOK
    152.32 NOK / 1000mL
    Save 37.25 NOK
    19% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Bestellinformation
ForUseWithApplication
Spezifikation
Product Type Anti-microbial Washing Lotion
Form Lotion
For Use With (Application) For antimicrobial body cleansing (in case of MRSA/ORSA, VRE) including shampooing. Through selected ingredients and the alcohol-free formulation the product possesses a considerably better skin compatibility than conventional decontamination preparations.