Learn More
Bochem™ Steel Zincked Rod
Description
- With thread M10
- Other sizes available
- 12 D/O
- Steel zincked
Specifications
Specifications
| Material | Steel Zincked |
| Length (Metric) | 600 mm |
| Diameter (Metric) Rod | 12 mm |
| Includes | M10 thread |
| Product Type | Steel Zincked Rod |
| Thread Size | M10 |
| For Use With (Equipment) | For stand bases M10 |
| Quantity | 1 Ea. |