Learn More
Bochem™ Steel Stand Base
Product Code. 10411203
Change view
Click to view available options
Length (Metric):
180 mm
210 mm
300 mm
315 mm
Width (Metric):
100 mm
130 mm
150 mm
200 mm
Weight (Metric):
0.8 kg
1.3 kg
2.2 kg
3 kg
Unit Size:
Each
Product Code. Length (Metric) Width (Metric) Weight (Metric) unitSize
10411203 315 mm 200 mm 3 kg Each
10199780 180 mm 100 mm 0.8 kg Each
10644711 210 mm 130 mm 1.3 kg Each
10038990 300 mm 150 mm 2.2 kg Each
4 options
Product Code. 10411203
Brand: Bochem™ 5034
Only null left
Add to basket
Add to basket
Manufactured according to DIN 12892. Bochem™ Steel Stand Base features M10 thread.
  • Steel
  • Manufactured according to DIN 12892
  • M10 thread
  • One adjustable foot
  • Varnished
Specifications
Material Steel
Height (Metric) 6 mm
Length (Metric) 315 mm
Width (Metric) 200 mm
Includes M10 thread
Product Type Stand Base
Thread Size M10
Color Green
Quantity 1 Ea.
Weight (Metric) 3 kg