Learn More
Bochem™ Steel Stand Base
Description
- Steel
- Manufactured according to DIN 12892
- M10 thread
- One adjustable foot
- Varnished
Specifications
Specifications
| Material | Steel |
| Height (Metric) | 6 mm |
| Length (Metric) | 180 mm |
| Width (Metric) | 100 mm |
| Includes | M10 thread |
| Product Type | Stand Base |
| Thread Size | M10 |
| Color | Green |
| Quantity | 1 Ea. |
| Weight (Metric) | 0.8 kg |