Learn More
Bochem™ Steel Rods for Stand Base
Description
- With thread M10
- 1500mm in length, 12mm in diameter
Specifications
Specifications
| Material | Steel |
| Length (Metric) | 1500 mm |
| Diameter (Metric) Outer | 12 mm |
| Product Type | Rods for Stand Bases |
| Quantity | 1 Pk. |