Learn More
Bochem™ Steel Opening Tong
Product Code. p-8006750
Change view
Click to view available options
Diameter (Metric) Outer:
8 mm
20 mm
Unit Size:
Each
Product Code. Diameter (Metric) Outer unitSize
10346101 20 mm Each
10729451 8 mm Each
2 options
Product Code. 10346101
Brand: Bochem™ 12968
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Made from 18/10 stainless steel. The Bochem Steel Opening Tong can open different sized caps.
  • Opening tong
  • For aluminum caps
  • Made from 18/10 stainless steel
Specifications
Diameter (Metric) Outer 20 mm
Material Steel