Learn More
asecos™ Steel Disposal Bin
Product Code. 17866486 Shop All asecos Products
Change view
Click to view available options
Capacity (Metric):
20 L
35 L
50 L
65 L
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
17866486 20 L Each
17876546 35 L Each
17836496 50 L Each
17856426 65 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17866486 Supplier asecos™ Supplier No. 39379
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
The Steel Disposal Bin is ideal for the safe collection of solvent-soaked cleaning rags and other waste.
  • Sheet Steel
  • Disposal bin ideal for safe disgard of solvant-soaked rags.
  • Self-closing lid with damping, large, ergonomically shaped handle on the rear of the container for easy emptying of the container
  • Opening by large pedal with anti-slip surface
  • Available in various capacities.
Specifications
Capacity (Metric) 20 L
Depth (English) 15.04 in.
Depth (Metric) 382.00 mm
Capacity (English) 5.3 gal.
Color Red
Height (Metric) 429.00 mm
Weight (English) 12.35 lb.
Weight (Metric) 5.6 kg
Width (Metric) 385.00 mm
Width (English) 15.16 in.
Height (English) 16.89 in.
Material Steel
Product Type Liquid Disposal Can