Learn More
Brady™ Maximum Chain Security
Product Code. 11746925
Click to view available options
Diameter (Metric):
8 mm
10 mm
Unit Size:
1 pack
Pack of 2
Product Code. 11746925
Brand: Brady™ 805846
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
More secure than steel cables.
  • Supplied with a fabric sleeve which will protect paintwork and chrome from damage
Specifications
Product Type Chain
Alternate Part Number CHAIN SECURITY 10 MM 2 PK
Diameter (Metric) 10 mm
Quantity 2 Pk.