missing translation for 'onlineSavingsMsg'
Learn More

Stearic Acid, Sigma-Aldrich™

Product Code. 15888370
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15888370 1 kg 1kg
15878370 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15888370 Supplier Sigma Aldrich Supplier No. 8006731000
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-11-4

Specifications

Melting Point 68°C to 70°C
Boiling Point 370°C/1013 hPa
Molecular Formula CH3(CH2)16COOH
MDL Number MFCD00002752
Quantity 1 kg
Synonym Stearic acid, Stearic acid
Molecular Weight (g/mol) 284.48
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.