Learn More
Microspec™ Steam Sterilisation Indicator Strips
Product Code. 15702185
Change view
Click to view available options
Quantity:
250
Unit Size:
Pack of 250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15702185 250 Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15702185 Supplier Microspec™ Supplier No. X1
  • ONLINE EXCLUSIVE PRICE
    1805.00 NOK
    1672.08 NOK / Pack of 250
    Save 132.92 NOK
    7% Off
Estimated Shipment: 17-12-2026
Predicts completion of autoclave cycle
  • Quickly ascertain whether an autoclave cycle has completed
Specifications
Autoclavable Autoclavable
Color White
For Use With (Application) Sterilization indication
Product Type Steam Sterilization Strip
Temperature (Metric) 121°C
Net Weight (English) 6.24 oz.
Net Weight (Metric) 177 g
Quantity 250
Width (English) Tray 3.15 in.
Width (Metric) Tray 80 mm
Sterility Sterile
Dimensions (L x W x H) 165 x 78 x 26 mm