Learn More
Invitrogen™ Stealth RNAi™ siRNA Negative Control Lo GC
Click to view available options
Quantity:
250 μL
Unit Size:
Each
Description
Stealth RNAi™ siRNA Negative Controls are a great way to measure the effect of your experimental Stealth RNAi™ siRNA duplex versus background. These controls are: –Designed in 3 levels of GC content (Lo, Med, Hi) with 3 sequences available per level so researchers can match GC content to their experimental Stealth RNAi™ siRNA –Not homologous to anything in vertebrate transcriptome –Tested not to induce stress response Stealth RNAi™ siRNA Negative Control Kit contains all three controls (Lo, Med, Hi GC content-excluding sequences #2 and #3) and each is also available separately. Get the ideal negative controls for your RNAi experiment so you can assess your results accurately.
Specifications
Specifications
| Content And Storage | Supplied at 250 μl at 20 μM. Store at -20°C. Guaranteed stable for 6 months when properly stored. |
| Control Type | Negative Control |
| RNAi Type | siRNA |
| For Use With (Application) | RNAi, Transfection |
| Label or Dye | Unconjugated |
| Product Line | Stealth RNAi™ siRNA |
| Product Type | siRNA |
| Quantity | 250 μL |
For Research Use Only. Not for use in diagnostic procedures.
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido