Learn More
Brady™ Static Dissipative Thermal Transfer Printable Labels
Product Code. 11583664
Click to view available options
Height (Metric):
5.1 mm
6.4 mm
7 mm
9.5 mm
11.1 mm
12.7 mm
Vertical Repeat (Metric):
7.6 mm
8.9 mm
9.5 mm
12.1 mm
13.6 mm
15.2 mm
Width (Metric):
6.4 mm
9.5 mm
12.7 mm
13.8 mm
16.5 mm
17.8 mm
19.1 mm
22.9 mm
25.4 mm
31.8 mm
38.1 mm
50.8 mm
Labels per Row:
1
2
3
4
5
7
12
Unit Size:
1 pack
1 roll
Product Code. 11583664
Brand: Brady™ 029912
Only null left
Add to basket
Add to basket
Printable, static dissipative polyester labels for Thermal Transfer Printers.
  • Material is UL and CSA approved with its respective ribbon
  • Ideal for Circuit Board Labeling
  • For smooth surfaces
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance UL Recognized, CSA Accepted, Meets surface resistivity values in the recommended range for dissipative ESD packaging materials as defined by ANSI/ESD S541-2008 (between 10 and 10 ohms), Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Gloss
Format Die-cut
Material Static-dissipative Polyester
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Circuit Board Labeling, Electrical Labeling, General Identification
For Use With (Equipment) BBP81, i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500
Horizontal Repeat (Metric) 8.1 mm
Labels per Row 7
Min. Operating Temperature (Metric) -40°C
Model THT-70-473-20
No. per Pack 20000 Label(s) / Roll
Quantity 20,000 Labels
Shape Square
Size 6.35 mm (W) x 6.35 mm (H)
Temperature Range (English) -40°F to 356°F
Temperature Range (Metric) -40°C to +120°C
Thickness (English) 0.0034 in.
Thickness (Metric) 0.09 mm
Vertical Repeat (Metric) 8.9 mm
Dimensions (LxW) 6.35 mm (W) x 6.35 mm (H)
Length (English) 0.25 in.
Width (English) 0.25 in.
Width (Metric) 6.4 mm
Width (English) Printable Area 6.35 mm
Width (Metric) Printable Area 6.4 mm
Height (English) 0.25 in.
Height (Metric) 6.4 mm
Height (Metric) Printable Area 6.4 mm
Alternate Part Number THT-70-473-20