missing translation for 'onlineSavingsMsg'
Learn More

STAT5B Mouse anti-Human, Clone: 1D9B11, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66427-1-IG-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16848295

  • 3828.53 NOK / 150µL
Estimated Shipment: 16-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein mediates the signal transduction triggered by various cell ligands, such as IL2, IL4, CSF1, and different growth hormones. It has been shown to be involved in diverse biological processes, such as TCR signaling, apoptosis, adult mammary gland development, and sexual dimorphism of liver gene expression. This gene was found to fuse to retinoic acid receptor-alpha (RARA) gene in a small subset of acute promyelocytic leukemias (APLL). The dysregulation of the signaling pathways mediated by this protein may be the cause of the APLL.
Specifications

Specifications

STAT5B
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P51692
STAT5B
STAT5B Fusion Protein Ag2709
150 μL
Primary
Human
Antibody
IgG2a
Western Blot
1D9B11
Unconjugated
STAT5B
STAT5, STAT5B
Mouse
Protein A
RUO
6777
-20°C
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers