missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT5b Antibody (STAT5B/2657), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08628AF594
This item is not returnable.
View return policy
Description
STAT5b Monoclonal antibody specifically detects STAT5b in Human samples. It is validated for ELISA, Protein Array
Specifications
| STAT5b | |
| Monoclonal | |
| Alexa Fluor 594 | |
| signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B | |
| Recombinant full-length human STAT5b protein (Uniprot: P51692) | |
| 100 μg | |
| Signal Transduction | |
| 6777 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Protein Array | |
| STAT5B/2657 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction