missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13028-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The STAT 4 protein is a transcription factor activated by hormone and cytokines. STAT 4 is most similar to STAT 1 (52%) and STAT 3 (47%). Functionally, STAT 4 is similar to other STAT family members in that it can be tyrosine phosphorylated by JAK 1 or JAK 2. STAT 4 forms homodimers and heterodimers with related STAT family members. Tyrosine phosphorylated STAT 4 can bind the interferon g activated site (GAS), and serine phosphorylation is also required for maximal transcriptional activity. STAT 4 expression is restricted to the thymus, spleen, and testis.Specifications
STAT4 | |
Polyclonal | |
Unconjugated | |
STAT4 | |
SLEB11, STAT4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20849, 367264, 6775 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P42228, Q14765 | |
Stat4 | |
STAT4 Fusion Protein Ag3660 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |