Learn More
Corning™ Starter Sacrificial Inks for Matribot™ Bioprinter
Product Code. 17780829 Shop All Corning Products
Click to view available options
:
Starter Sacrificial Ink; Quantity: 10 Syringes
Product Code. 17780829
Brand: Corning™ undefined
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Starter Sacrificial Inks are sacrificial material and inks for educational purposes. Designed for Use with Matribot™ Bioprinter.
  • Water-soluble gel used as a sacrificial material and ink for educational purposes
  • Ideal for evaluating the printing of complex structures, prior to printing with human cells
  • Designed for use with Corning™ Matribot™ Bioprinter
TRUSTED_SUSTAINABILITY
Specifications
Color Transparent Blue
For Use With (Equipment) Corning Matribot Bioprinter
Product Type Sacrificial Ink
Capacity (Metric) 2.7 mL/Syringe
Description Sterile-A Quality
Model Starter
For Use With (Application) Evaluating the printing of complex structures, prior to printing with human cells
Quantity 10 Syringes
Storage Requirements 4°C to 8°C