Learn More
Gibco™ Starter Pak No. 3
Product Code. 16279831 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16279831 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16279831 Supplier Gibco™ Supplier No. 215368
Make Your Process Development Count
Use promo code 27978
View all promotions
 Restricted Item
Estimated Shipment: 07-12-2026
Provides a variety of yeast and soy products, ideal for processes where an animal origin-free medium is preferred.
  • It provides conveniently packaged, 100-g, non-GMP samples of these peptones that are best suited for human and animal vaccine production and contains:
    • Bacto Yeast Extract and Gibco Yeast Extract—these peptones contain a mixture of peptides, amino acids, carbohydrates, and vitamins to support optimal growth in microbial species, and are ideal for the production of vaccines
    • Bacto TC Yeastolate—this peptone is ideal for CHO-based applications of biotherapeutic monoclonal antibodies and recombinant proteins, as well as vaccine applications
    • Phytone Peptone and Difco Soytone—these soy-based supplements provide a nutritious source of carbohydrates for successful use in microbial fermentation. Blending these soy peptones with yeast peptones has been shown to provide additional benefit to cultures.
    • Bacto Malt Extract—a water-soluble portion of malted barley, this peptone also provides carbohydrates for a variety of microbial fermentation processes
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.
Form Powder
Product Type Dehydrated Culture Media
For Use With (Application) Human and animal vaccine productions
Quantity 1 Ea.
Source Animal origin-free, yeast and soy