Learn More
SI Analytics™ Starter Kit KF for Titronic™ 500, TitroLine™ 7000, 7500, 7750, 7800
Shop All SI Analytics Products
Click to view available options
For Use With (Equipment):
Titronic 500, TitroLine 7000, 7500, 7750, 7800
Unit Size:
Each
Description
- Starter kit KF
- For use with Titronic 500, TitroLine 7000,7500, 7750, 7800
Specifications
Specifications
| Description | Starter kit KF |
| Includes | Molecular sieve, needles with syringes and glass wool |
| For Use With (Equipment) | Titronic 500, TitroLine 7000, 7500, 7750, 7800 |