Learn More
Heidolph™ StarFish MonoBlock
Product Code. 11791659 Shop All Heidolph Instruments Products
Change view
Click to view available options
Holds:
5 Flasks
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
11791659 5 Flasks Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11791659 Supplier Heidolph™ Supplier No. 5058000100
  • ONLINE EXCLUSIVE PRICE
    8090.00 NOK
    7950.08 NOK / Pack of 1
    Save 139.92 NOK
    1% Off
Estimated Shipment: 20-10-2026
Add to basket
Add to basket
Accommodate up to 5 flasks at your StarFish magnetic stirrer workstation. Heidolph™ StarFish MonoBlock, constructed of one piece, accepts inserts to hold variously sized flasks.
  • Accommodates up to 5 flasks.
  • Accepts inserts to hold various size flasks.
TRUSTED_SUSTAINABILITY
Specifications
Description Monoblock
For Use With (Equipment) Starfish Workstation, MR-Hei Magnetic Stirrers
Holds 5 Flasks