Learn More
Starch solution, Honeywell™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Specifications
Specifications
| Chemical Name or Material | Starch solution |
| Density | 1.15-1.25g/mL at 20°C |
| Quantity | 100 mL |