missing translation for 'onlineSavingsMsg'
Learn More

Starch, MP Biomedicals

Product Code. p-3545929 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 lb.
100 lb.
Unit Size:
100lb
25lb
This item is not returnable. View return policy

Product Code. 11495692

Brand: MP Biomedicals 0290295625

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Approx. 75% amylopectin, 25% amylose

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-84-9
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.297
InChI Key GUBGYTABKSRVRQ-ASMJPISFSA-N
Synonym alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
PubChem CID 439341
ChEBI CHEBI:18167
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O

Specifications

Quantity 25 lb.
Chemical Name or Material Starch, From Corn
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.