Learn More
Novacyt - Microgen Bioproducts Staph ID
Product Code. 17333221 Shop All Novacyt Group Products
Change view
Click to view available options
Quantity:
20 x 3 mL bottles, 20 Test strips
Unit Size:
20 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17333221 20 x 3 mL bottles, 20 Test strips 20 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17333221 Supplier Novacyt - Microgen Bioproducts Supplier No. MID69
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Suitable for use with clinical and veterinary samples.
Microgen™ Staph-ID has been developed for the identification of commonly encountered Staphylococcus spp. isolated from clinical and veterinary samples. These bacteria, although commensal, are recognised as potential human pathogens e.g. S. aureus and S. epidermidis. Coagulase negative Staphylococcus spp. are common colonisers of prosthetic valves and in dwelling catheters. Gram stain (positive), catalase (positive) and latex agglutination/coagulase tests are performed as pre-tests on the isolate. Colony pigmentation is recorded and then 1-2 colonies are inoculated into the suspending medium. The suspension is inoculated into the 12 well test strip and incubated at 37°C. After 24 hours incubation the strips are read and the Nitrate and PYR reagents added. The resulting 5 digit numerical code is entered in to the software and an identification returned.
As with all Microgen™ biochemical ID systems, Staph-ID is supported by the simplest-to-use and most feature-packed Computer Identification System Software (MID60) available, delivering the most comprehensive and up-to-date databases.
Species identified:
  • Staphylococcus spp.
  • S. aureus subsp. aureus
  • S. aureus subsp. anaerobius
  • S. auricularis
  • S. caprae
  • S. capitis subsp. capitis
  • S. capitis subsp. urealyticus
  • S. carnosus
  • S. chromogenes
  • S. cohnii subsp. conhii
  • S. cohnii subsp. urealyticum
  • S. epidermidis
  • S. haemolyticus
  • S. hominis subsp. hominis
  • S. hominis subsp. novobiosepticus
  • S. hyicus
  • S. intermedius
  • S. lentus
  • S. lugdenensis
  • S. saccharolyticus
  • S. saprophyticus
  • S. schleiferi subsp. schleiferi
  • S. schleiferi subsp. coagulans
  • S. sciuri
  • S. simulans
  • S. warneri
  • S. xylosus Kocuria spp.
  • K. kristinae
  • K. rosea
  • K. carniphila Kytococcus spp.
  • Ky. sedentarius Micrococcus spp.
  • M. luteus
  • M. lylae
Specifications
Certifications/Compliance ISO13485 ISO9001
Content And Storage 2°C to 8°C
CE Marker Yes
Clia Complexity Non-waived
Detectable Analytes Gelatin, Malonate, Inositol, Sorbitol, Rhamnose, Sucrose, Lactose, Arabinose, Adonitol, Raffinose, Salicin, Arginine, TDA, Nitrate
Format 20 x 3 mL bottles, 20 Test strips
Sample Type Bacterial Isolate
Sensitivity >99%
Type Test Strip
For Use With (Application) An Identification System for Staphylococci of Clinical Animal Health and Environmental Importance
Includes 20 Test strips
No. of Reactions 12 per test strip
Organism Group Monera
Packaging Type Carton with pouches and strip holding tray
Quantity 20 x 3 mL bottles, 20 Test strips
Species Staphylococci of clinical animal health and environmental importance
Specificity >99%
Target Organism Class Bacteria
Test Time 18 to 24 hours