Learn More
Stannous Chloride, Dihydrate, 98.0 to 101.0%, ACS, MilliporeSigma™
Product Code. 15865050
Change view
Click to view available options
Quantity:
2.5 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15865050 2.5 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15865050 Supplier Supelco Supplier No. 1078151000
 Restricted Item
Estimated Shipment: 26-11-2026
High-purity Tin(II) chloride dihydrate designed for precise qualitative and quantitative analytical applications.
TRUSTED_SUSTAINABILITY
Specifications
CAS 10025-69-1
Color White
Physical Form Crystalline Solid
Quantity 2.5 kg
Packaging Glass Bottle
Assay Percent Range 98.0 to 101.0%
Molecular Formula SnCl2·2H2O
Synonym Tin(II) Chloride, Dihydrate
Molecular Weight (g/mol) 225.65
Formula Weight 225.65
Grade Not Available
Chemical Name or Material Stannous Chloride, Dihydrate, For ACS analysis