Learn More
Fisherbrand™ Standing Rest on Glides with Footring
Product Code. 17776689 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Type: Chair
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17776689 Type: Chair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17776689
  • ONLINE EXCLUSIVE PRICE
    2200.00 NOK
    2152.52 NOK / Pack of 1
    Save 47.48 NOK
    2% Off
View all promotions
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
The most flexible support for sitting, standing and combined workplaces. This standing rest boasts a soft non-slip PU-foam upholstery, offering maximum safety and comfort. It offers a particular wide range of heights that can be adjusted steplessly.
  • Can be used as lab stool or standing rest
  • With integrated back support
  • 360° rotation of the seat
  • Particular wide range of heights: Lowest height: Serves as stool / Highest height: Serves as standing rest
  • Solid construction, extremely tough
  • 10 years guarantee
Specifications
Product Type Chair
Color Black
Adjustable (Height) 510 to 780 mm
Adjustable Height adjustable
Certifications/Compliance GS-mark for tested safety according to DIN 68877
Finish PU-foam
For Use With (Application) For standing work in the laboratory
Frame Material Aluminium, polished
Includes Seat, gas spring, gas spring cover, 5-star foot base, glides
Lift Type Gas lift height adjustment
Load Bearing Capacity (Metric) 120 kg
Seating Height Adjustable from 510 to 780 mm
Warranty 10 years
Weight (Metric) 13 kg
Wheel Type Glides
Width (Metric) 640 mm
With Backrest Yes
Footrest Yes
With Lumbar Support Yes
Seating Type High Bench Height
Show More Show Less
Meets biosafety level S1 and S2, GMP compliant