Learn More
Tenak™ Standard Rack for Upright Freezer, 100 mm Box Height
Standard Rack fits into any upright freezer and can be used with all standard cryoboxes.
1075.00 NOK - 1390.00 NOK
Specifications
Material | Stainless Steel |
---|---|
For Use With (Equipment) | Upright Freezers |
Height (Metric) | 100 mm (Box) |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
15230704
|
Tenak™
TE23212 |
Boxes (12) |
1250.00 NOK
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
12439500
|
Tenak™
TE23198 |
Boxes (4) |
1100.00 NOK
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
15210704
|
Tenak™
TE23210 |
Boxes (8) |
1155.00 NOK
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
15270694
|
Tenak™
TE23200 |
Boxes (6) |
1075.00 NOK
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
15270704
|
Tenak™
TE23222 |
Boxes (15) |
1390.00 NOK
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
15290694
|
Tenak™
TE23202 |
Boxes (9) |
1155.00 NOK
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
Description
- Measure the inside height and depth of your freezer when choosing the right rack
- Racks can be used with all freezer brands
- Constructed of stainless steel
Specifications
Stainless Steel | |
Upright Freezers | |
100 mm (Box) |