Learn More
Tenak™ Standard Rack for Microtiterplates, for Upright Freezers, 16 mm Shelf Height
Stainlesss Steel Standard Rack for use in upright freezers is designed to hold microtiterplates.
Brand:  Tenak™ TE27450
Additional Details : Weight : 3.40000kg
 View more versions of this product
Product Code. 15290924
  • 2400.00 NOK / Each
Estimated Shipment: 02-08-2024
Description
  • For use in upright freezers
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • Constructed of stainless steel
Specifications
Specifications
Stainless Steel
Microtiterplates (45)
450 x 135 x 153 mm
Upright Freezers
9 x 5
16 mm