Learn More
Tenak™ Standard Rack for Microtiterplates, for Upright Freezers, 16 mm Shelf Height
Stainlesss Steel Standard Rack for use in upright freezers is designed to hold microtiterplates.
Brand: Tenak™ TE27526
413.96 NOK valid until 2024-05-31
Use promo code "21638" to get your promotional price.
Additional Details : Weight : 6.60000kg
Description
- For use in upright freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of stainless steel
Specifications
Stainless Steel | |
Microtiterplates (114) | |
540 x 135 x 323 mm |
Upright Freezers | |
19 x 6 | |
16 mm |