Learn More
Tenak™ Standard Rack for Microtiterplates, for Chest Freezers, 25 mm Shelf Height
Stainlesss Steel Standard Rack for chest freezers is designed to hold microtiterplates.
Brand:  Tenak™ TE27136
Additional Details : Weight : 1.45000kg
 View more versions of this product
Product Code. 15210884
  • 1515.00 NOK / Each
Estimated Shipment: 19-07-2024
Description
  • For use in chest freezers
  • Racks come with a locking rod to secure the cryoboxes
  • Constructed of stainless steel
Specifications
Specifications
Locking rod
Stainless Steel
135 x 92 x 455 mm
Microtiterplates (17)
Chest Frseezers
25 mm