Learn More
Sampling Systems™ Hand Reel EX
Product Code. 15899341 Shop All Sampling Systems Products
Change view
Click to view available options
Diameter (Metric):
175 mm
Unit Size:
Each
Product Code. Diameter (Metric) unitSize
15899341 175 mm Each
1 options
Product Code. 15899341
Brand: Sampling Systems™ 5070D05
Only null left
Add to basket
Add to basket
Sampling Systems™ Standard Heavy Duty Ex Hand Reel is for use with the Sub Sampler Pro.
  • For use with Sub Sampler Pro
  • Polypropylene construction
Specifications
Product Type Hand Reel
Material Conductive Polyamide, Aluminum
Diameter (Metric) 175 mm
Width (Metric) 55 mm