Learn More
Koettermann™ Standard Hanging Cabinet, 1200x480x366mm
Koettermann™ Standard Hanging Cabinet, 1200x480x366mm provides suitable storage to accommodate a variety of laboratory needs.
6094.74 NOK - 6655.10 NOK
Specifications
Dimensions (D x W x H) | 366 x 1200 x 480 mm |
---|---|
No. of Doors | 2 |
No. of Shelves | 1 |
Product Type | Wall Mounted Cabinet |
Product Code | Brand | Locking Mechanism | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Locking Mechanism | Price | Quantity & Availability | |||||
15723082
|
Koettermann™
307-00166 |
Yes |
6655.10 NOK
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
15713082
|
Koettermann™
307-00165 |
No |
6094.74 NOK
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
Description
- Wall-mounted cabinet
- Two doors
- One shelf
- Lockable
- Side panels
Specifications
366 x 1200 x 480 mm | |
2 | |
1 | |
Wall Mounted Cabinet |