Learn More
Kinematica Standard Dispersing Aggregate for PT 2500 E
Product Code. 11980439 Shop All Kinematica Products
Click to view available options
Diameter (Metric) Probe:
12 mm
25 mm
Volume Range (Metric):
100 mL to 2 L
100 mL to 2.5 L
Speed:
21 m/s
26 m/s
Unit Size:
Each
Product Code. 11980439
Brand: Kinematica 11030069
Only null left
Add to basket
Add to basket
Uses two rows of teeth, is suitable for a large spectrum of laboratory applications, and is reliable and efficient.
  • Selection and applications:
    • All standard dispersing tasks
    • Grinding of animal and human tissue samples in a variety of fluids and volumes
    • Sample preparation for the extraction and dissolution of organic materials
    • Sample preparation for medical diagnostic procedures as well as DNA analysis
    • Extractions of systems / substances from vegetable-derived samples
    • Disruption of animal and human cells
    • Deagglomeration of united cell structures
    • Intensive mixing
Specifications
Product Type Standard Dispersing Aggregate
Speed 21 m/s
Volume Range (Metric) 100 mL to 2 L
Diameter (Metric) Probe 12 mm
Length (Metric) 192 mm
Model PT-DA 20/2EC-E19
Packaging Quantity 1 Ea.