Learn More
Koettermann™ Standard Chemical Cabinet, 1200x720x516mm
Product Code. 14885371
Click to view available options
No. of Doors:
2
Unit Size:
Each
Product Code. 14885371
Brand: Koettermann™ 37500020
This item requires a price update prior to being ordered.
Request a Price for this item here.
Koettermann™ Standard Chemical Cabinet, 1200x720x516mm provides suitable storage to accommodate a variety of laboratory needs, but is especially suited for storing chemicals.
  • Chemical cabinet
  • Two doors
  • One shelf
  • Lockable
Specifications
Dimensions (D x W x H) 516 x 1200 x 720 mm
Includes Lock
No. of Doors 2
No. of Shelves 1
Product Type Chemical Cabinet