Learn More
Standard Buffer Solution pH 6.87, WTW™
Description
- As standard buffering solutions, the buffers pH 6.865 and PL 7 are available in 250 ml bottles
- PTB/NIST traceable standard buffer solutions (work reference buffer solutions) for precision applications - safe use
- They have a high accuracy to ± 0.02 pH
- Standard buffering solutions as per DIN 19266-01: 2000
- Universal buffering solutions
Specifications
Specifications
| Boiling Point | 100°C |
| Color | Colorless |
| Certificate of Analysis | DIN/NIST |
| For Use With (Application) | Precision applications |
| pH | 6.87 |
| Physical Form | Liquid |
| Quantity | 250 mL |
| Packaging | Bottle |