Learn More
Heidolph™ Stand S2 XXL
Product Code. 10562805 Shop All Heidolph Instruments Products
Change view
Click to view available options
Length (Metric):
1000 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
10562805 1000 mm Each
1 options
Product Code. 10562805
Brand: Heidolph™ 5701220000
Only null left
Add to basket
Add to basket
A unique stand for our entire overhead stirrer series.
  • A unique stand for our entire overhead stirrer series.
TRUSTED_SUSTAINABILITY
Specifications
Length (English) 39.4 in.
Length (English) Base 14.6 in.
Length (Metric) 1000 mm
Length (Metric) Base 370 mm
Diameter (English) Rod 1 in.
Diameter (Metric) Rod 25 mm
Product Type Stand
Weight (English) 13.2 lb.
Weight (Metric) 6 kg