Learn More
Bochem™ Stainless Steel Z Shape Test Tube Stands
Product Code. 10010811
Change view
Click to view available options
Dimensions (L x W):
50 x 85 mm
50 x 110 mm
50 x 220 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
10010811 50 x 110 mm Each
10234081 50 x 220 mm Each
10616191 50 x 85 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10010811 Supplier Bochem™ Supplier No. 8293
Estimated Shipment: 17-02-2027
Specifications
Holds Tubes (17 mm)
Material Stainless Steel (18/10)
Positions 25
Dimensions (L x W) 50 x 110 mm
Diameter (Metric) Opening 17 mm
Product Type Test Tube Rack