Learn More
Bochem™ Stainless Steel Z Shape Test Tube Stands
Product Code. 10616191
Change view
Click to view available options
Dimensions (L x W):
50 x 85 mm
50 x 110 mm
50 x 220 mm
Unit Size:
Each
Product Code. Dimensions (L x W) unitSize
10616191 50 x 85 mm Each
10010811 50 x 110 mm Each
10234081 50 x 220 mm Each
3 options
Product Code. 10616191
Brand: Bochem™ 8292
Only null left
Specifications
Holds Tubes (13 mm)
Material Stainless Steel (18/10)
Positions 25
Dimensions (L x W) 50 x 85 mm
Diameter (Metric) Opening 13 mm
Diameter (Metric) Outer 13 mm
Product Type Test Tube Rack