Learn More
Buerkle™ Worm-Threaded Hose Clips
Product Code. 16616703 Shop All Bürkle Products
Click to view available options
Tubing Size:
10 to 16 mm dia.
12 to 20 mm dia.
16 to 27 mm dia.
20 to 32 mm dia.
25 to 40 mm
8 to 12 mm dia.
Unit Size:
Pack of 10
Product Code. 16616703
Brand: Buerkle™ 86772540
Only null left
Add to basket
Add to basket
Manufactured from stainless steel 1.4016.
  • With screw thread
  • Screw is zinc-coated
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 25 to 40 mm
Material Stainless Steel
For Use With Hoses
Packaging Quantity 10 Pk.
Tubing Size 25 to 40 mm
Product Type Worm-threaded Hose Clip