Learn More
Bochem™ Stainless Steel Wire Baskets
Product Code. 10603652
Change view
Click to view available options
Height (Metric):
150 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
10603652 150 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10603652 Supplier Bochem™ Supplier No. 10028
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Estimated Shipment: 09-12-2026
Specifications
Material 18/10 E-Poli
Dimensions (L x W x H) 180 x 180 x 150 mm
Length (Metric) 180 mm
Height (Metric) 150 mm
Width (Metric) 180 mm