Learn More
Bochem™ Stainless Steel Wire Basket, Square
Description
- Stainless steel
- Square
- Flat bottom for stability
Specifications
Specifications
| Dimensions (L x W) | 150 L x150 Wmm |
| Dimensions (L x W x H) | 150 x 150 x 150 mm |
| Product Type | Wire Basket |